Learn More
Abeba™ Anatom 2268 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand: Abeba™ 2268/37
Additional Details : Weight : 1.40000kg
Description
- CE EN ISO 20345:2011 S3 SRC
- Smooth leather
- Black
- Steel toe cap
- Shock absorbant PU midsole
- Metalfree penetration resistance
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
Specifications
CE EN ISO 20345:2011 S3 SRC | |
Leather | |
Slip-resistant | |
2268 |
Black | |
37 | |
Polyurethane | |
Steel |