Learn More
Abeba™ Anatom 2280 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 2280/44
Additional Details : Weight : 1.88000kg
 View more versions of this product
Product Code. 15608097
  • 140.57€ / 1 pair
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S3 SRC
  • Microfiber
  • White
  • Steel toe cap
  • Shock absorbant PU midsole
  • Metalfree penetration resistance
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • Antistatic
  • Cold insulating lining
  • Adjustable instep area
  • velcro fastening
  • Reflective material
  • suitable for heel and foot lift
  • cushioned ankle support
Specifications
Specifications
CE EN ISO 20345:2011 S3 SRC
Microfiber
Slip-resistant
2280
White
44
Polyurethane
Steel