Learn More
Abeba™ Anatom 2280 Shoes
Product Code. 15628087
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15628087
Brand: Abeba™ 2280/36
Only null left
Add to basket
Add to basket
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S3 SRC
  • Microfiber
  • White
  • Steel toe cap
  • Shock absorbant PU midsole
  • Metalfree penetration resistance
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • Antistatic
  • Cold insulating lining
  • Adjustable instep area
  • velcro fastening
  • Reflective material
  • suitable for heel and foot lift
  • cushioned ankle support
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S3 SRC
Color White
Material Microfiber
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 2280
Toe Material Steel