Learn More
Abeba™ Anatom 2615 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand: Abeba™ 2615/43
Additional Details : Weight : 1.26600kg
Description
- CE EN ISO 20345:2011 S1 SRC
- Microfiber, perforated
- Black
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Adjustable instep area
- Double velcro fastening
- Reflective material
- Suitable for heel and foot lift
Specifications
CE EN ISO 20345:2011 S1 SRC | |
Microfiber | |
Slip-resistant | |
2615 |
Black | |
43 | |
Polyurethane | |
Steel |