Learn More
Abeba™ Anatom 32168 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Marke: Abeba™ 32168/40
Weitere Details : Weight : 1.34000kg
Beschreibung
- CE EN ISO 20345:2011 S2 SRC
- Smooth leather
- Black
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Scratch-resistant coating in heel and toe area
Spezifikation
CE EN ISO 20345:2011 S2 SRC | |
Leather | |
Slip-resistant | |
32168 |
Black | |
40 | |
Polyurethane | |
Steel |