Learn More
Abeba™ Anatom 32169 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 32169/44
Additional Details : Weight : 1.40000kg
 View more versions of this product
Product Code. 15629427
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • CE EN ISO 20345:2011 S2 SRC
  • Smooth leather
  • Black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Low cut
  • Discontinued model
  • Scratch-resistant coating in heel and toe area
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRC
Leather
Slip-resistant
32169
Black
44
Polyurethane
Steel