Learn More
Abeba™ Anatom 32171 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 32171/38
Additional Details : Weight : 1.30000kg
 View more versions of this product
Product Code. 15699427
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Suitable for hygienically demanding areas
  • Suitable for heel and foot lift
  • Scratch-resistant coating in heel and toe area
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRC
Microfiber
Slip-resistant
32171
Black
38
Polyurethane
Steel