Learn More
Abeba™ Anatom 32172 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 32172/41
Additional Details : Weight : 1.30000kg
 View more versions of this product
Product Code. 15659447
  • 128.73€ / 1 pair
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber
  • White
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Suitable for hygienically demanding areas
  • Suitable for heel and foot lift
  • Scratch-resistant coating in heel and toe area
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRC
Microfiber
Slip-resistant
32172
White
41
Polyurethane
Steel