Learn More
Abeba™ Anatom 32189 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Marque: Abeba™ 32189/43
Informations supplémentaires : Weight : 1.18800kg
Description
- CE EN ISO 20345:2011 S1 SRC
- Velours with breathable textile inlays
- Black
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Adjustable instep area
- velcro fastening
- Reflective material
- Suitable for heel and foot lift
Spécification
CE EN ISO 20345:2011 S1 SRC | |
Velour | |
Slip-resistant | |
32189 |
Black | |
43 | |
Polyurethane | |
Steel |