Learn More
Abeba™ Anatom 32243 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 32243/36
Additional Details : Weight : 1.44300kg
 View more versions of this product
Product Code. 15659507
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20345:2011 S1P SRC
  • Smooth leather with breathable textile inlays
  • Black
  • ATEX design
  • Steel toe cap
  • Shock absorbant PU midsole
  • Metalfree penetration resistance
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
Specifications
Specifications
CE EN ISO 20345:2011 S1P SRC
Leather
Slip-resistant
32243
Black
36
Polyurethane
Steel