Learn More
Abeba™ Anatom 32270 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 32270/43
Additional Details : Weight : 1.52400kg
 View more versions of this product
Product Code. 15609597
  • 135.00€ / 1 pair
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S3 SRC
  • Smooth leather
  • Black in ATEX design
  • Steel toe cap
  • Shock absorbant PU midsole
  • Metalfree penetration resistance
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Scratch-resistant coating in toe and heel area
Specifications
Specifications
CE EN ISO 20345:2011 S3 SRC
Leather
Slip-resistant
32270
Black
43
Polyurethane
Steel