Learn More
Abeba™ Anatom 32280 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand: Abeba™ 32280/46
Additional Details : Weight : 1.88000kg
Description
- CE EN ISO 20345:2011 S3 SRC
- Microfiber
- White
- Steel toe cap
- Shock absorbant PU midsole
- Metalfree penetration resistance
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Cold insulating lining
- Adjustable instep area
- velcro fastening
- Reflective material
- suitable for heel and foot lift
- cushioned ankle support
Specifications
CE EN ISO 20345:2011 S3 SRC | |
Microfiber | |
Slip-resistant | |
32280 |
White | |
46 | |
Polyurethane | |
Steel |