Learn More
ABEBA™ Anatom Black Safety Boots S3
Product Code. 16368071
Change view
Click to view available options
Size:
49
50
51
52
Unit Size:
1 pair
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16368071 49 1 pair
16378071 50 1 pair
16388071 51 1 pair
16398071 52 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16368071 Supplier ABEBA™ Supplier No. 32268/49
Only null left
Add to basket
Add to basket
ABEBA™ Anatom Black Safety Boots S3 made of water resistant smooth leather with a slip resistant PU2D outsole.
  • Comfort, robustness, durability
  • Comply to EN ISO 20345 : 2012, S3, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU double density outsole
  • black water resistant smooth leather
  • Steel toe cap (200 joules)
  • Textile antipuncture midsole
  • Scratch-resistant coating in toe and heel area
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • Removable ACC Wave insole
Specifications
Certifications/Compliance CE EN ISO 20345:2012 S3 SRC ESD
Closure Type Laces
Color Black
Eyelets Material Stainless steel
Liner Material Breathable silver ions lining
Material Leather
Size 49
Sole Features Slip and abrasion resistant, ESD dissipative, shock absorbent
Sole Material Polyurethane2D
Style Mid cut boots
Toe Material Steel
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) EN ISO 20345 : 2012; DIN EN 61340
Resistant To Water
Waterproof Yes