Learn More
Emma Safety Footwear Andes XD Safety Shoes
Features a steel toe cap and an integrated stainless steel plate. The double-layer, anti-slip PU sole provides extra protection from hazards on the ground.
Brand:  Emma Safety Footwear MM30556844
Additional Details : Weight : 1.69500kg
 View more versions of this product
Product Code. 15968331
  • 124.02€ / 1 pair
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
Andes XD
Oiled Nubuck
44