Learn More
Emma Safety Footwear Andes XD Safety Shoes
Product Code. 15928331
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Product Code. 15928331
Brand: Emma Safety Footwear MM30556840
Only null left
Add to basket
Add to basket
Features a steel toe cap and an integrated stainless steel plate. The double-layer, anti-slip PU sole provides extra protection from hazards on the ground.
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Design Andes XD
Material Oiled Nubuck
Size 40