Learn More
Fisherbrand™ Anemometer Helix
Product Code. 11901107 Shop All Fisherbrand Products
Click to view available options
Airflow Rate:
0.5 to 40 m/sec.
Unit Size:
Each
Product Code. 11901107
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Type Anemometer Helix
Accuracy ±2%
Airflow Rate 0.5 to 40 m/sec.
Height (Metric) 180 mm
Length (Metric) 72 mm
Power Supply Details 9V battery
Precision ±2%
Scale m/s, ft/min, km/h, knots and mile/h
Temperature (Metric) -10 to +50°C
Weight (Metric) 700 g