Learn More
Sampling Systems™ Angular Beaker
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
2 L
Material:
Polypropylene
Stainless Steel
Unit Size:
Each
Description
- Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
- A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
- Attachments are interchangeable and simply click into position
Specifications
Specifications
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Capacity (Metric) | 1 L |
| Disposable | Reusable |
| Product Type | Swabbing Template |
| Material | Polypropylene |