missing translation for 'onlineSavingsMsg'
Learn More

Anhydrotetracycline Hydrochloride, TRC

Product Code. 30338157
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Product Code. 30338157

Brand: Toronto Research Chemicals TRCA65605050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Anhydrotetracycline hydrochloride
CAS 13803-65-1
Purity Grade Notes HPLC
Molecular Formula C22H22N2O7.HCl
Quantity 50 mg
Synonym Anhydrotetracycline Hydrochloride, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-, monohydrochloride, (4S,4aS,12aS)- (9CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-, monohydrochloride (8CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-, monohydrochloride, [4S-(4íÅ1⁄2±,4aíÅ1⁄2±,12aíÅ1⁄2±)]-, Chlortetracycline Hydrochloride Impurity J as Hydrochloride, Tetracycline Hydrochloride Impurity C as Hydrochloride, Chlortetracycline Impurity J as Hydrochloride, Tetracycline Impurity C as Hydrochloride
InChI Formula InChI=1S/C22H22N2O7.ClH/c1-8-9-5-4-6-12(25)13(9)17(26)14-10(8)7-11-16(24(2)3)18(27)15(21(23)30)20(29)22(11,31)19(14)28;/h4-6,11,16,25-27,31H,7H2,1-3H3,(H2,23,30);1H/t11-,16-,22-;/m0./s1
SMILES Cl.CN(C)[C@H]1[C@@H]2Cc3c(C)c4cccc(O)c4c(O)c3C(=O)[C@]2(O)C(=O)C(=C1O)C(=O)N
IUPAC Name (4S,4aS,12aS)-4-(dimethylamino)-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 462.88
Formula Weight 462.1194
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.