Learn More
Laboratoires Anios™ Aniosafe Savon Doux HF
Laboratoires Anios™ Aniosafe Savon Doux HF is a gentle soap recommended for frequent hand hygiene and general body cleansing in hospital environments.
85.50€ - 176.00€
Specifications
Product Type | Gentle Soap |
---|---|
Certifications/Compliance | NF EN ISO 11930 |
Composition | Water, Disodium Lauroamphodiacetate, Sodium Lauryl Sulfate, Hexylene Glycol, Glycerin, Linoleamide DEA, Disodium Ricinoleamido EASulfosuccinate, Benzyl Alcohol, Sodium Benzoate, PEG-200 Hydrogenated Glyceryl Palmate and PEG-7 Glyceryl Cocoate, Parfum (Fragrance), Lactic acid. |
For Use With (Application) | General body cleansing in hospital environments |
Packaging | Bottles with screw pump |
Product Code | Brand | Packaging | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging | Volume (Metric) | Price | Quantity & Availability | ||||
15591813
|
Laboratoires Anios™
1918228VB |
Bottles with screw pump | 500 mL |
85.50€
Pack of 12 |
In Stock
Log in to see stock. |
||||
15585290
|
Laboratoires Anios™
1918229VB |
Bottles with screw pump | 1 L |
176.00€
Pack of 12 |
In Stock
Log in to see stock. |
||||
Description
- Dermatologically tested
- Enriched with moisturizer
- Preservation of the product demonstrated according to NF EN ISO 11930
Specifications
Gentle Soap | |
Water, Disodium Lauroamphodiacetate, Sodium Lauryl Sulfate, Hexylene Glycol, Glycerin, Linoleamide DEA, Disodium Ricinoleamido EASulfosuccinate, Benzyl Alcohol, Sodium Benzoate, PEG-200 Hydrogenated Glyceryl Palmate and PEG-7 Glyceryl Cocoate, Parfum (Fragrance), Lactic acid. | |
Bottles with screw pump |
NF EN ISO 11930 | |
General body cleansing in hospital environments | |
12 Bottles |