Learn More
Laboratoires Anios™ Aniosgel 85 NPC
Product Code. 15600115
Click to view available options
Volume (Metric):
500 mL
1 L
Packaging:
Airless bottles
Pump Bottles
Unit Size:
Pack of 12
Product Code. 15600115
Brand: Laboratoires Anios™ 1644763
Laboratoires Anios™ Aniosgel 85 NPC contains no allergenic substances, fragrance, or coloring, and is suitable for hygienic treatment and surgical disinfection of hands
  • No allergenic substances, fragrance, or coloring
  • Broad antimicrobial spectrum
  • Hygienic treatment in 30 sec.
  • For hygienic treatment and surgical disinfection
  • Active against bacteria, mycobacteria, yeasts, molds, viruses
Specifications
Product Type Sterilizer/Disinfectant Hand Gel
Certifications/Compliance (Europe) EN 13727, EN 13727, EN 14348, EN 13624, EN 14476, EN 14476, EN 1500, EN 12791
Composition Ethanol in the presence of thickening, moisturizing and emollient agents, and water.
For Use With (Application) Hygienic treatment and surgical disinfection of hands
Packaging Pump Bottles
Quantity 12 Bottles
Volume (Metric) 1 L
compliance-icons compliance-icons
Product Identifier
  • ANIOSGEL 85 NPC
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403-Store in a well-ventilated place.
Supplemental information
  • MIXTURE LIST-Contains : ETHYL ALCOHOL, PROPAN-2-OL