missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Annexin V Binding Buffer
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Description
- Annexin V Binding Buffer is an isotonic buffer used in conjunction with Annexin V dyes (e.g. Catalog #100-0329 and either 7-AAD (Catalog #75001) or Propidium Iodide (Catalog #75002))
- This buffer contains an optimal concentration of calcium that facilitates the binding of Annexin V to phosphatidylserine on the cell surface, which is essential for the detection of early-stage cell apoptosis by flow cytometry
Specifications
Specifications
| Quantity | 50 mL |
| For Use With (Application) | Flow Cytometry |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction