Learn More
Agencinox™ Anodised Aluminum Colored Sterilization Boxes
Use these boxes for a variety of general laboratory applications. Agencinox™ Anodised Aluminum Colored Sterilization Boxes keep instruments safe and secure during sterilization process.
Brand:  Agencinox™ 8036
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11822692
  • 43.96€ / Each
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
  • Anodized aluminum colored instrument boxes
  • Available in a variety of sizes
Specifications
Specifications
Natural
215 mm
50 mm
Colored Sterilization Box
100 mm