missing translation for 'onlineSavingsMsg'
Learn More

Agencinox™ Anodised Aluminum Colored Sterilization Boxes

Use these boxes for a variety of general laboratory applications. Agencinox™ Anodised Aluminum Colored Sterilization Boxes keep instruments safe and secure during sterilization process.

Brand:  Agencinox™ 8036R

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11852682

  • 51.59€ / Each
Estimated Shipment: 16-05-2024
to see stock.
Add to basket

Description

Description

  • Anodized aluminum colored instrument boxes
  • Available in a variety of sizes
Specifications

Specifications

Red
Rectangle
215 mm
50 mm
Aluminum
Sterilization Box
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions