missing translation for 'onlineSavingsMsg'
Learn More

6x-His Tag Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA1983B

383.80 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.



Additional Details : Weight : 0.01000kg

Product Code. 11598711

  • 404.00€ / 200µg
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PA1-983B detects 6x-His Epitope tagged proteins PA1-983B has been successfully used in Western blot procedures. The PA1-983B immunogen is a synthetic peptide corresponding to residues H H H H H H.

6x-His Tag Polyclonal antibody specifically detects 6x-His Tag in Tag samples. It is validated for Immunocytochemistry, Western Blot.
Specifications

Specifications

6x-His Tag
Polyclonal
Unconjugated
6His tag; 6X His; His Tag; Histidine Tag
Synthetic peptide corresponding to residues H H H H H H.
200 μg
Primary
Antibody
IgG
Immunocytochemistry, Western Blot
2 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Rabbit
Antigen affinity chromatography
RUO
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions