missing translation for 'onlineSavingsMsg'
Learn More

Myc Tag Monoclonal Antibody, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen R95025

245.33 EUR valid until 2024-03-29
Use promo code "21653" to get your promotional price.



Additional Details : Weight : 0.02000kg

Product Code. 10206792

  • 263.00€ / Each
Estimated Shipment: 19-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NOTE: R950-25 was previously sold under product #46-0603. When re-ordering, please use #R950-25. This antibody recognizes amino acid sequence: Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu-. This product contains enough material for 25 Western blots. Western blot against 500 ng of recombinant protein gave a strong signal using chemiluminescent or alkaline phosphatase detection reagents. For Western blot, dilute in PBS containing 0.05% Tween-20 and 5% nonfat, dry milk (PBSTM).

Myc Tag Monoclonal antibody specifically detects Myc Tag in Tag samples. It is validated for ELISA, Western Blot.
Specifications

Specifications

Myc Tag
Monoclonal
Unconjugated
MYC
46-0603; AU016757; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; MRTL; MYC; Myc protein; Myc proto-oncogene protein; Myc Tag; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; Niard; Nird; Proto-oncogene c-Myc; R95025; R950-25; transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog
Mouse
purified
RUO
4609
Antibody
IgG1
ELISA, Western Blot
∽1 mg/mL
PBS with 0.01% sodium azide
P01106
MYC
c-Myc synthetic peptide: Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu-
50 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions