Learn More
C1orf57 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530852
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (80%), Rhesus Monkey (97%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C1orf57 Polyclonal antibody specifically detects C1orf57 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
C1orf57 | |
Polyclonal | |
Unconjugated | |
Ntpcr | |
2310079N02Rik; AI449709; C1orf57; cancer-related nucleoside-triphosphatase; Cancer-related nucleoside-triphosphatase homolog; CH1073-266P11.1; HCR-NTPase; NTPase; Ntpcr; Nucleoside triphosphate phosphohydrolase; nucleoside-triphosphatase C1orf57 homolog; nucleoside-triphosphatase, cancer-related; RGD1306192; RP4-659I19.2; RP4-678E16.2; zgc:92420 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84284 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9BSD7 | |
Ntpcr | |
Recombinant fragment corresponding to a region within amino acids 1 and 179 of Human C1orf57 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |