missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathelicidin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSC00009W
Additional Details : Weight : 0.01000kg
Description
Glycerol (1:1) may be added for additional stability. Reconstitute with 150 μL of distilled water. OSC00009W can be used with blocking peptide GSC00002P.
Cathelicidin antimicrobial protein is an antimicrobial protein found in specific granules of polymorphonuclear leukocytes. Expressed in bone marrow and testis and neutrophils.Specifications
Cathelicidin | |
Polyclonal | |
CAMP | |
CAP-18, hCAP-18, antibacterial protein LL-37, CAMP, CRAMP, FALL39, hCAP18 | |
Rabbit | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
P49913 | |
CAMP | |
A synthetic peptide from human Cathelicidin antimicrobial peptide | |
RUO | |
820 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |