missing translation for 'onlineSavingsMsg'
Learn More

CD1e Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA526941

Additional Details : Weight : 0.01000kg

Product Code. 13200959

  • 473.00€ / 100µg
Estimated Shipment: 10-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD1e Polyclonal antibody specifically detects CD1e in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

CD1e
Polyclonal
PBS with 0.09% sodium azide
P15812
CD1E
KLH conjugated synthetic peptide between 184-212 amino acids from the central region of human CD1E
400 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CD1E
CD1A; CD1e; CD1E antigen, e polypeptide; CD1e molecule; differentiation antigen CD1-alpha-3; hCD1e; leukocyte differentiation antigen; R2; R2G1; sCD1e; T-cell surface glycoprotein CD1e, membrane-associated; T-cell surface glycoprotein CD1e, soluble; thymocyte antigen CD1E
Rabbit
Antigen affinity chromatography, Protein A
RUO
913
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions