missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1 Monoclonal Antibody (1H4), PerCP, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110250
Additional Details : Weight : 0.01000kg
Description
ICAM-1 Monoclonal antibody specifically detects ICAM-1 in Human samples. It is validated for Flow Cytometry.Specifications
ICAM-1 | |
Monoclonal | |
PerCP | |
ICAM1 | |
BB2; CD54; cell surface glycoprotein P3.58; Human rhinovirus receptor; ICAM; Icam1; ICAM-1; ICAM-1; CD54 homolog; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; intercellular adhesion molecule-1; intercellular adhesion molecule-1 precursor; Ly 47; Ly-47; major group rhinovirus receptor; MALA2; MALA-2; MyD10; P3.58; sCD54; soluble CD54; Surface antigen of activated B cells | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
3383 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
1H4 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P05362 | |
ICAM1 | |
Raji cells and spleen cells fused with NS1 cells | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b |