Learn More
CEACAM1, Human, Matched Antibody Pair (AP43), Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00000634-AP43.1Set
Product Code. 16131784
  • 843.00€ / 1 set
Includes: Capture antibody: mouse monoclonal anti-CEACAM1 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-CEACAM1, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
CEACAM1
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CEACAM1.
BGP/BGP1/BGPI
Mouse
RUO
634
Human
Liquid
ELISA
Biotin
CEACAM1
CEACAM1
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CEACAM1 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-CEACAM1, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.