Learn More
Cholera Toxin beta Polyclonal Antibody, HRP, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA173189
Additional Details : Weight : 0.01000kg
Description
The PA1-73189 antibody reacts with Cholera toxin beta subunit. PA1-73189 has been successfully used in ELISA applications. The PA1-73189 antibody was raised against purified Choleragenoid.
Cholera Toxin beta Polyclonal antibody specifically detects Cholera Toxin beta in Bacteria samples. It is validated for ELISA.Specifications
Cholera Toxin beta | |
Polyclonal | |
HRP | |
CTB; CTX; VC1456 | |
Purified Choleragenoid. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
ELISA | |
1-2 mg/mL | |
PBS with 10mg/mL BSA and 0.002% thimerosal | |
Rabbit | |
purified | |
RUO | |
Bacteria | |
Antibody | |
IgG |