Learn More
anti-CLPS, Human, Matched Antibody Pair, Abnova™
Product Code. 16161894
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
Product Code. Quantity unitSize
16161894 1 Set 1 set
1 options
Product Code. 16161894
Brand: Abnova H00001208AP41.1Set
Includes
Capture antibody: mouse monoclonal anti-CLPS, IgG2a Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-CLPS, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Specifications
Antigen CLPS
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Biotin
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CLPS.
Gene CLPS
Gene Symbols CLPS
Host Species Mouse
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1208
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CLPS, IgG2a Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-CLPS, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid