missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523448
Additional Details : Weight : 0.01000kg
Description
DLL3 Polyclonal antibody specifically detects DLL3 in Human, Mouse, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
DLL3 | |
Polyclonal | |
Unconjugated | |
Dll3 | |
delta like canonical Notch ligand 3; Delta3; delta-like 3; delta-like 3 (Drosophila); delta-like protein 3; Dll3; Drosophila Delta homolog 3; M-Delta-3; pu; pudgy; SCDO1 | |
Rabbit | |
Protein A | |
RUO | |
10683, 13389 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
O88516, Q9NYJ7 | |
Dll3 | |
A portion of amino acid 100 to 150 of human DLL3 | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG |