missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUX4 Monoclonal Antibody (P4H2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516147
Additional Details : Weight : 0.10000kg
Description
DUX4 Monoclonal antibody specifically detects DUX4 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
DUX4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9UBX2 | |
Dux4 | |
Synthetic peptide corresponding to the C-terminal of human DUX4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
P4H2 | |
Unconjugated | |
Dux4 | |
AW822073; double homeobox; double homeobox 4; double homeobox 4-like; double homeobox B-like 1; double homeobox protein 10; Double homeobox protein 4; double homeobox protein 4/10; Dux; DUX10; Dux4; DUX4L; Duxbl; Duxbl1; EG664783; homeobox protein; RGD1311053 | |
Mouse | |
Protein G | |
RUO | |
100288687 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |