missing translation for 'onlineSavingsMsg'
Learn More

anti-F2, Human, Matched Antibody Pair, Abnova™

Product Code. 16182004
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
Product Code. Quantity unitSize
16182004 1 Set 1 set
1 options
This item is not returnable. View return policy

Product Code. 16182004

Brand: Abnova H00002147AP52.1Set

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Capture antibody: mouse monoclonal anti-F2, IgG Mix Kappa (300μg) , Detection antibody: rabbit polyclonal anti-F2 (50μL) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.

To detect and quantify protein level of human

Specifications

Antigen F2
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human F2.
Gene F2
Gene Alias PT
Gene Symbols F2
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2147
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-F2, IgG Mix Kappa (300 ug) 2. Detection antibody: rabbit polyclonal anti-F2 (50 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.