Learn More
Merck Millipore anti-GABA A Receptor α1 ; Polyclonal
Brand:  Merck Millipore AB5592-200UL
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15124017
  • 1135.00€ / 200µL
Estimated Shipment
20-05-2021
Add to basket
Description
Description
Immunohistochemistry, Western Blotting
Specifications
Specifications
GABA-A Receptor α1
Affinity Purified
Peptide corresponding to amino acids 1-16 from mouse or rat GABA(A) α1 subunit (Swiss-Prot accession number P18504, residues 28-43 of the precursor).
RUO
Primary
Rat
Unconjugated
Rabbit
200μL
Neuroscience