Learn More
Merck Millipore anti-GABA A Receptor α1 ; Polyclonal
Brand: Merck Millipore AB5592-200UL
Additional Details : Weight : 1.00000kg
Description
Immunohistochemistry, Western Blotting
Specifications
GABA-A Receptor α1 | |
Affinity Purified | |
Peptide corresponding to amino acids 1-16 from mouse or rat GABA(A) α1 subunit (Swiss-Prot accession number P18504, residues 28-43 of the precursor). | |
RUO | |
Primary | |
Rat |
Unconjugated | |
Rabbit | |
200μL | |
Neuroscience | |
Polyclonal |