missing translation for 'onlineSavingsMsg'
Learn More

GAS8 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA543123

Additional Details : Weight : 0.00010kg

Product Code. 15555882

  • 472.00€ / 100µL
Estimated Shipment: 18-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Peptide sequence: RALKVELKEQ ELASEVVVKN LRLKHTEEIT RMRNDFERQV REIEAKYDKK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%

GAS8 Polyclonal antibody specifically detects GAS8 in Human samples. It is validated for Immunocytochemistry, Western Blot.
Specifications

Specifications

GAS8
Polyclonal
Unconjugated
GAS8
beta growth arrest specific protein 11; CILD33; DRC4; Dynein regulatory complex subunit 4; Gas11; GAS-11; Gas8; GAS-8; growth arrest specific 11; growth arrest specific 8; growth arrest specific protein 11; growth arrest-specific protein 11; Growth arrest-specific protein 8; MGC138326
Rabbit
Affinity Chromatography
RUO
2622
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
O95995
GAS8
Synthetic peptide directed towards the middle region of human GAS8
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions