Learn More
GFAP, Human, Matched Antibody Pair (AP52), Abnova™
Product Code. 16172094
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
Product Code. Quantity unitSize
16172094 1 Set 1 set
1 options
Denne vare kan ikke returneres. Se returpolitik
Produktkode 16172094
missing translation for 'mfr': Abnova H00002670AP52.1Set
Includes
Capture antibody: mouse monoclonal anti-GFAP, IgG2a Kappa (100μg) , Detection antibody: rabbit purified polyclonal anti-GFAP (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Specifications
Antigen GFAP
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human GFAP.
Gene GFAP
Gene Alias FLJ45472
Gene Symbols GFAP
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2670
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-GFAP, IgG2a Kappa (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-GFAP (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid