missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GUSB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526357
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with porcine based on sequence homology.
GUSB Polyclonal antibody specifically detects GUSB in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
GUSB | |
Polyclonal | |
Unconjugated | |
GUSB | |
Ac2-223; adipose storage deficiency; AI747421; asd; beta glucuronidase; beta-D-glucuronidase; beta-G1; Beta-glucuronidase; BG; g; glucuronidase beta; glucuronidase, beta; Gur; Gus; Gusb; Gus-r; Gus-s; Gus-t; Gus-u; Gut; lysosomal enzyme; MPS7; si:ch211-160e1.7; si:ct573103.7 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
2990 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P08236 | |
GUSB | |
KLH conjugated synthetic peptide between 335-362 amino acids from the central region of human GUSB | |
400 μL | |
Primary | |
Human | |
Antibody | |
Ig |