Learn More
Hepatitis B Virus Surface Ayw/Adw Monoclonal Antibody (HB5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119264
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes the following Hepatitis B virus (HBV) subtypes: ayw2, ayw3, ayw4, ayr, adw2, adw4, adrq+, adrq-. Specificity of the antibody HB5 was verified by ELISA on panel of virus subtypes. The antibody HB5 does not cross-block with the antibody HB3 (MA1-19263).
Hepatitis B Virus Surface Ayw/Adw Monoclonal antibody specifically detects Hepatitis B Virus Surface Ayw/Adw in Virus samples. It is validated for ELISA, Immunocytochemistry.Specifications
Hepatitis B Virus Surface Ayw/Adw | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Mouse | |
sequential chromatography | |
RUO | |
Virus | |
Antibody | |
IgG2a |
ELISA, Immunocytochemistry | |
HB5 | |
Unconjugated | |
HBsAg; HBV; hepatitis b virus surface antigen; HepB | |
Purified HBsAg from human plasma. | |
100 μg | |
Primary | |
4° C, do not freeze | |
Liquid |