missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537170
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 30 kDa. Purity is >95% by SDS-PAGE.
HOXC12 Polyclonal antibody specifically detects HOXC12 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
HOXC12 | |
Polyclonal | |
Unconjugated | |
HOXC12 | |
HOC3F; homeo box C12; homeobox C12; homeobox protein Hox-3.8; Homeobox protein Hox-3F; Homeobox protein Hox-C12; Homeobox protein Hox-C12 (Hox-3F); HOX3; Hox-3.8; HOX3F; HOXC12; Hoxc-12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15421, 300262, 3228 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P31275, Q8K5B8 | |
HOXC12 | |
A synthetic peptide corresponding to residues in human HOX-C12 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |