missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Human IgG (Gamma chain) Cross-Adsorbed Secondary Antibody, HRP, Invitrogen™

Goat Polyclonal Secondary Antibody

Brand:  Invitrogen 628420

Additional Details : Weight : 0.50000kg


Product Code. 10691234

  • 259.00€ / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Human IgG (Gamma chain) Cross-Adsorbed Polyclonal antibody specifically detects Human IgG (Gamma chain) Cross-Adsorbed in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Specifications

Specifications

Human IgG (Gamma chain) Cross-Adsorbed
Polyclonal
HRP
Goat
Affinity chromatography
RUO
Human
Liquid
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1 mg/mL
PBS with 40% glycerol, 4mg/mL BSA and 0.19% Kathon™ CG; pH 7.4
Gamma Immunoglobin
1 mL
Secondary
4° C
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Goat anti-Human IgG (Gamma chain) Cross-Adsorbed Secondary Antibody, HRP

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : glycerin, 5-Chloro-2-methyl-3(2H)-isothiazolone

SDS
Documents

Documents

Certificates
Promotions

Promotions