missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV Type 16 E7 Monoclonal Antibody (716-325), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA123087
Additional Details : Weight : 0.01000kg
Description
MA1-23087 detects HPV-16 in Virus samples.
HPV Type 16 E7 Monoclonal antibody specifically detects HPV Type 16 E7 in Virus samples. It is validated for ELISA, Western Blot.Specifications
HPV Type 16 E7 | |
Monoclonal | |
5.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG2a |
ELISA, Western Blot | |
716-325 | |
Unconjugated | |
HPV 16-E7; HPV16E7; human papillomavirus | |
Human papilloma virus oncoprotein E7, Type 16. | |
250 μg | |
Primary | |
4° C, do not freeze | |
Liquid |