missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG1,κ, Clone: R3-34, BD

Brand:  BD Biosciences 562868

Product Code. 15892309

  • ONLINE EXCLUSIVE PRICE
    292.00€
    291.04€ / Each
    Save 0.96€
    0.33% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Following immunization of a rat with mouse immunoglobulin (Ig), the Ig from the R3-34 hybridoma was identified as a non-reactive clone. The R3-34 immunoglobulin was selected as an Ig isotype control following screening for low background staining on a variety of mouse and human cells and tissues.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon™ BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon™ BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.

Host Species: Rat
Clone: R3-34
Isotype: IgG1, κ
Immunogen: Mouse immunoglobulin

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flow Cytometry
R3-34
Brilliant Violet 421
Rat
Affinity Purified
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse immunoglobulin
50 μg
Isotype Control (Primary)
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.