Learn More
anti-IgG2a κ, Rat, PE-Cy™7, Clone: R35-95, Isotype Control, BD
Brand: BD Biosciences 552784
Additional Details : Weight : 0.01000kg
Description
The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: R35-95
Isotype: IgG2a κ
Immunogen: Mouse Pooled Immunoglobulin
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
0.2mg/mL | |
Rat | |
Affinity Purified | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
R35-95 | |
PE-Cyanine7 | |
Mouse Pooled Immunoglobulin | |
0.1 mg | |
IgG2a κ |