missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCAT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522965
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: antigen standard for LCAT (transient overexpression lysate), Lipoprotein fraction of whole mouse plasma.
LCAT Polyclonal antibody specifically detects LCAT in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
LCAT | |
Polyclonal | |
Unconjugated | |
LCAT | |
AI046659; D8Wsu61e; Lcat; lecithin cholesterol acyltransferase; lecithin-cholesterol acyltransferase; lecithin-cholesterol acyltransferase Lcat; phosphatidylcholine-sterol acyltransferase; phosphatidylcholine--sterol O-acyltransferase; Phospholipid-cholesterol acyltransferase; testicular secretory protein Li 24 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
P04180, P16301 | |
LCAT | |
Carboxy-terminal histidine-tagged mouse recombinant LCAT purified from the medium of CHO cells overexpressing mouse LCAT cDNA. | |
RUO | |
16816, 3931 | |
-20°C or -80°C if preferred | |
Liquid |