Learn More
CD85j (ILT2) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529567
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Human (98%), Rhesus Monkey (83%), Chimpanzee (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD85j (ILT2) Polyclonal antibody specifically detects CD85j (ILT2) in Human samples. It is validated for Western Blot.Specifications
CD85j (ILT2) | |
Polyclonal | |
Unconjugated | |
LILRB1 | |
CD85 antigen-like family member J; CD85j; Ig-like transcript 2; ILT2; ILT-2; Immunoglobulin-like transcript 2; leucocyte Ig-like receptor B1; leukocyte immunoglobulin like receptor B1; leukocyte immunoglobulin-like receptor subfamily B member 1; leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform; leukocyte immunoglobulin-like receptor subfamily B member 3-like; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1; LILRB1; LIR1; LIR-1; LOC100912456; MIR7; MIR-7; Monocyte/macrophage immunoglobulin-like receptor 7; myeloid inhibitory receptor 7; PIRB; PIR-B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10859 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8NHL6 | |
LILRB1 | |
Recombinant fragment corresponding to a region within amino acids 88 and 353 of Human LILRB1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |