missing translation for 'onlineSavingsMsg'
Learn More

anti-MAP2K6, GADD45A Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0487.1Set

Product Code. 16191404

  • 677.00€ / 1 set

Please to purchase this item. Need a web account? Register with us today!


Includes: MAP2K6 rabbit purified polyclonal antibody (20μg); GADD45A mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
This item is not returnable. View return policy

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the MAP2K6 protein, and the other against the GADD45A protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

MAP2K6
Monoclonal/Polyclonal
MAP2K6
MAP2K6
1 Set
Primary
Antibody pair set content: 1. MAP2K6 rabbit purified polyclonal antibody (20 ug) 2. GADD45A mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
In situ PLA
Unconjugated
MAPKK6/MEK6/MKK6/PRKMK6/SAPKK3
Mouse, Rabbit
RUO
5608/1647
Human
MAP2K6 & GADD45A Protein Protein Interaction Antibody Pair
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.