missing translation for 'onlineSavingsMsg'
Learn More

CSF1R Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA514569

Additional Details : Weight : 0.01000kg

Product Code. 10598730

  • 516.00€ / 100µg
Estimated Shipment: 12-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CSF1R Polyclonal antibody specifically detects CSF1R in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

CSF1R
Polyclonal
Unconjugated
Csf1r
AI323359; CD115; CD115 antigen; C-FMS; colony stimulating factor 1 receptor; colony stimulating factor 1 receptor, formerly McDonough feline sarcoma viral (v-fms) oncogene homolog; CSF-1 receptor; CSF1R; CSF-1R; CSF-1-R; CSFIR; Csfmr; CSFR; EC 2.7.10.1; FIM2; Fim-2; FMS; fms proto-oncogene; HDLS; kinase CSFR; macrophage colony stimulating factor I receptor; macrophage colony-stimulating factor 1 receptor; McDonough feline sarcoma viral (v-fms) oncogene homolog; M-CSFR; M-CSF-R; mrfms; OTTHUMP00000224093; protein-tyrosine kinase; proto-oncogene c-Fms; proto-oncogene fms
Rabbit
Affinity chromatography, Protein A
RUO
1436
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
P07333
Csf1r
KLH conjugated synthetic peptide between 940-971 amino acids from the C-terminal region of human MCSF Receptor (CSF1R)
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions