missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555, Invitrogen™

Goat Polyclonal Secondary Antibody

Brand:  Invitrogen A21426

244.15 EUR valid until 2024-03-29
Use promo code "21653" to get your promotional price.



Additional Details : Weight : 0.00450kg

Product Code. 10318122

  • 257.00€ / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Product will be shipped at Room Temperature.

Mouse IgM (Heavy chain) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgM (Heavy chain) Cross-Adsorbed in Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.
Specifications

Specifications

Mouse IgM (Heavy chain) Cross-Adsorbed
Polyclonal
Alexa Fluor 555
Goat
purified
RUO
Mouse
Liquid
Immunocytochemistry, Immunohistochemistry, Western Blot
2 mg/mL
PBS with 5mM sodium azide; pH 7.5
Mouse Mu immunoglobulin
500 μg
Secondary
4° C, store in dark
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions